siRNA Details
| Gene Symbol: NUP43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198887.2 | NP_942590.1 | 8 | 1029 | ||
| NR_104456.1 | 8 | 1029 | ||||
| XM_005266960.4 | XP_005267017.1 | 8 | 2259 | |||
| XM_005266961.3 | XP_005267018.1 | 9 | 1079 | |||
| XM_005266962.3 | XP_005267019.1 | 8 | 3299 | |||
| XM_011535799.2 | XP_011534101.1 | 8 | 2341 | |||
| GenBank mRNA | AF514997.1 | - | 8 | 962 | ||
| AK023349.1 | - | 3 | 216 | |||
| AK074311.1 | - | 985 | ||||
| AK095994.1 | - | 8 | 985 | |||
| AK304244.1 | - | 6 | 673 | |||
| BC040121.1 | - | 6 | 674 | |||
| BC046637.1 | - | 1 | 777 | |||
| BC047539.1 | - | 1 | 1938 | |||
| BC065028.1 | - | 8 | 955 | 6042215 | ||
| BX647097.1 | - | 8 | 979 | |||

NUP43
Genomic Map