siRNA Details
| Gene Symbol: ZNF713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182633.2 | NP_872439.2 | 7 | 1985 | ||
| GenBank mRNA | AK097282.1 | - | 4 | 998 | ||
| BC094796.1 | - | 7 | 1807 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF713| Gene Symbol: ZNF713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182633.2 | NP_872439.2 | 7 | 1985 | ||
| GenBank mRNA | AK097282.1 | - | 4 | 998 | ||
| BC094796.1 | - | 7 | 1807 | |||