siRNA Details
Gene Symbol: ZNF713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182633.2 | NP_872439.2 | 7 | 1368 | ||
GenBank mRNA | AK097282.1 | - | 4 | 381 | ||
BC094796.1 | - | 7 | 1190 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182633.2 | NP_872439.2 | 7 | 1368 | ||
GenBank mRNA | AK097282.1 | - | 4 | 381 | ||
BC094796.1 | - | 7 | 1190 |