siRNA Details
| Gene Symbol: WDR86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284260.1 | NP_001271189.1 | 6 | 2002 | ||
| NM_001284261.1 | NP_001271190.1 | 5 | 1303 | |||
| NM_001284262.1 | NP_001271191.1 | 6 | 1439 | |||
| NM_198285.2 | NP_938026.2 | 6 | 1938 | |||
| XM_005249989.3 | XP_005250046.1 | 5 | 1799 | |||
| XM_011516148.1 | XP_011514450.1 | 6 | 1487 | |||
| XM_011516150.1 | XP_011514452.1 | 7 | 1691 | |||
| GenBank mRNA | AK125347.1 | - | 5 | 1252 | ||
| AK296049.1 | - | 6 | 1595 | |||
| BC107125.2 | - | 5 | 1335 | 40023951 | ||

WDR86
Genomic Map