siRNA Details
| Gene Symbol: RTN4RL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178570.2 | NP_848665.1 | 2 | 624 | ||
| GenBank mRNA | AF532858.1 | - | 2 | 295 | ||
| AX751514.1 | - | 2 | 295 | |||
| AY250221.1 | - | 2 | 295 | |||
| BC113673.1 | - | 2 | 317 | 8322725 | ||
| BC117276.1 | - | 2 | 317 | 40125827 | ||
| BC143818.1 | - | 2 | 317 | 9052333 | ||
| DQ864979.1 | - | 2 | 295 | |||

RTN4RL2
Genomic Map