siRNA Details
Gene Symbol: RAET1E-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW241754.1 | - | 1 | 262 |
Gene Symbol: RAET1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001788.3 | NP_001001788.2 | 3 | 567 | ||
NR_130110.1 | 3 | 567 | ||||
XM_011535800.2 | XP_011534102.1 | 3 | 3738 | |||
XM_017010831.1 | XP_016866320.1 | 3 | 3738 | |||
GenBank mRNA | AK310157.1 | - | 3 | 536 | ||
AY172579.1 | - | 3 | 493 | |||
AY172580.1 | - | 3 | 425 | |||
BC096719.1 | - | 3 | 425 | 40003404 | ||
BC098152.1 | - | 3 | 425 | 40003403 |
