siRNA Details
| Gene Symbol: LCE3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178435.3 | NP_848522.1 | 2 | 398 | ||
| XM_011509484.1 | XP_011507786.1 | 2 | 494 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LCE3E| Gene Symbol: LCE3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178435.3 | NP_848522.1 | 2 | 398 | ||
| XM_011509484.1 | XP_011507786.1 | 2 | 494 | |||