siRNA Details
| Gene Symbol: TBC1D26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178571.4 | NP_848666.2 | 14, 15 | 1333 | ||
| GenBank mRNA | BC028414.1 | - | 11, 12 | 3571 | ||
| BC047400.1 | - | 14, 15 | 1327 | 5297645 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBC1D26| Gene Symbol: TBC1D26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178571.4 | NP_848666.2 | 14, 15 | 1333 | ||
| GenBank mRNA | BC028414.1 | - | 11, 12 | 3571 | ||
| BC047400.1 | - | 14, 15 | 1327 | 5297645 | ||