siRNA Details
| Gene Symbol: TAS2R42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181429.2 | NP_852094.2 | 1 | 103 | ||
| GenBank mRNA | BC131742.1 | - | 1 | 103 | 40115195 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAS2R42| Gene Symbol: TAS2R42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181429.2 | NP_852094.2 | 1 | 103 | ||
| GenBank mRNA | BC131742.1 | - | 1 | 103 | 40115195 | |