siRNA Details
| Gene Symbol: KRT26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181539.4 | NP_853517.2 | 1 | 438 | ||
| GenBank mRNA | AJ564205.2 | - | 1 | 438 | ||
| BC132951.1 | - | 1 | 429 | 40146973 | ||
| BC136896.1 | - | 1 | 429 | 9020887 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT26| Gene Symbol: KRT26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181539.4 | NP_853517.2 | 1 | 438 | ||
| GenBank mRNA | AJ564205.2 | - | 1 | 438 | ||
| BC132951.1 | - | 1 | 429 | 40146973 | ||
| BC136896.1 | - | 1 | 429 | 9020887 | ||