siRNA Details
| Gene Symbol: SPATA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181727.1 | NP_859078.1 | 2 | 959 | ||
| XM_011533676.2 | XP_011531978.1 | 4 | 1505 | |||
| GenBank mRNA | AY221117.1 | - | 2 | 959 | ||
| BC033337.1 | - | 2 | 734 | 4838903 | ||
| BC126274.1 | - | 2 | 888 | 8991990 | ||
| BC136498.1 | - | 2 | 888 | 9020487 | ||

SPATA12
Genomic Map