siRNA Details
| Gene Symbol: GPR141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181791.1 | NP_861456.1 | 1 | 73 | ||
| XM_011515374.2 | XP_011513676.1 | 7 | 844 | |||
| XM_011515375.2 | XP_011513677.1 | 6 | 807 | |||
| XM_011515383.2 | XP_011513685.1 | 2 | 173 | |||
| XM_011515385.2 | XP_011513687.1 | 3 | 6024 | |||
| XM_011515386.2 | XP_011513688.1 | 3 | 246 | |||
| XM_011515388.2 | XP_011513690.1 | 2 | 392 | |||
| XM_011515389.2 | XP_011513691.1 | 2 | 431 | |||
| XM_017012160.1 | XP_016867649.1 | 5 | 340 | |||
| XM_017012161.1 | XP_016867650.1 | 5 | 594 | |||
| XM_017012162.1 | XP_016867651.1 | 6 | 1643 | |||
| XM_017012163.1 | XP_016867652.1 | 5 | 1607 | |||
| XM_017012164.1 | XP_016867653.1 | 2 | 181 | |||
| XM_017012165.1 | XP_016867654.1 | 3 | 6260 | |||
| XM_017012166.1 | XP_016867655.1 | 4 | 1563 | |||
| XM_017012167.1 | XP_016867656.1 | 3 | 273 | |||
| GenBank mRNA | AY288420.1 | - | 1 | 73 | ||
| BC120951.2 | - | 1 | 73 | 40115152 | ||
| BC120952.2 | - | 1 | 73 | 40115153 | ||
More Information
Set Membership: |
|

GPR141
Genomic Map