siRNA Details
| Gene Symbol: ZNF233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207005.1 | NP_001193934.1 | 5 | 858 | ||
| NM_181756.2 | NP_861421.2 | 5 | 888 | |||
| XM_017026758.1 | XP_016882247.1 | 5 | 1260 | |||
| XM_017026759.1 | XP_016882248.1 | 5 | 1041 | |||
| GenBank mRNA | AK055310.1 | - | 5 | 1041 | ||
| AK095351.1 | - | 5 | 879 | |||
| BC136734.1 | - | 4 | 773 | 9020724 | ||
| BC136735.1 | - | 4 | 773 | 9020725 | ||

ZNF233
Genomic Map