siRNA Details
| Gene Symbol: FAM101B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182705.2 | NP_874364.1 | 2 | 1262 | ||
| GenBank mRNA | AF085987.1 | - | 1 | 523 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM101B| Gene Symbol: FAM101B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182705.2 | NP_874364.1 | 2 | 1262 | ||
| GenBank mRNA | AF085987.1 | - | 1 | 523 | ||