siRNA Details
| Gene Symbol: TMPRSS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182973.1 | NP_892018.1 | 6, 7 | 739 | ||
| XM_011527978.2 | XP_011526280.1 | 8, 9 | 1102 | |||
| GenBank mRNA | AB109390.1 | - | 7, 8 | 856 | ||
| AJ488946.1 | - | 6, 7 | 739 | |||
| AJ488947.1 | - | 6, 7 | 739 | |||

TMPRSS9
Genomic Map