siRNA Details
| Gene Symbol: NHLRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198514.3 | NP_940916.2 | 11 | 2365 | ||
| GenBank mRNA | AK090631.1 | - | 11 | 2336 | ||
| AK098487.1 | - | 9 | 1479 | |||
| AL832090.1 | - | 2 | 336 | |||
| BC032598.1 | - | 7 | 1129 | 5529619 | ||
| BX538287.1 | - | 7 | 1159 | |||
| BX647641.1 | - | 9 | 1736 | |||

NHLRC2
Genomic Map