siRNA Details
| Gene Symbol: FAM111B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142703.1 | NP_001136175.1 | 3 | 1688 | ||
| NM_001142704.1 | NP_001136176.1 | 2 | 1643 | |||
| NM_198947.3 | NP_945185.1 | 4 | 1855 | |||
| GenBank mRNA | AK303520.1 | - | 3 | 1637 | ||
| AK304258.1 | - | 4 | 1803 | |||
| AY457926.1 | - | 4 | 1804 | |||
| BC130513.1 | - | 3 | 1745 | 40146544 | ||
| BC130539.1 | - | 3 | 1745 | 40146570 | ||

FAM111B
Genomic Map