siRNA Details
| Gene Symbol: TMEM179B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199337.2 | NP_955369.1 | 3 | 438 | ||
| XM_005273982.3 | XP_005274039.3 | 3 | 519 | |||
| GenBank mRNA | BC051355.1 | - | 3 | 413 | 6597282 | |
| BE439533.1 | - | 3 | 435 | |||

TMEM179B
Genomic Map