siRNA Details
| Gene Symbol: TEPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199046.2 | NP_950247.2 | 1 | 127 | ||
| NM_199456.2 | NP_955535.2 | 1 | 127 | |||
| GenBank mRNA | AY363222.1 | - | 1 | 127 | ||
| AY363223.1 | - | 1 | 127 | |||
| BC104458.1 | - | 1 | 118 | 40030655 | ||
| BC104459.1 | - | 1 | 118 | 40030657 | ||

TEPP
Genomic Map