siRNA Details
| Gene Symbol: C19orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198534.2 | NP_940936.2 | 7 | 1191 | ||
| XM_005272473.3 | XP_005272530.1 | 7 | 1362 | |||
| XM_011527993.1 | XP_011526295.1 | 7 | 1116 | |||
| XM_011527995.2 | XP_011526297.1 | 7 | 1191 | |||
| GenBank mRNA | AK093103.1 | - | 7 | 1176 | ||
| AK301623.1 | - | 2 | 152 | |||
| BC029824.1 | - | 7 | 1151 | 5170600 | ||
| BC034779.1 | - | 7 | 1140 | 5168945 | ||

C19orf45
Genomic Map