siRNA Details
| Gene Symbol: ZNF699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198535.2 | NP_940937.1 | 6 | 1589 | ||
| GenBank mRNA | AK095463.1 | - | 4 | 1101 | ||
| BC109267.1 | - | 5 | 1219 | 40025983 | ||
| BC109268.1 | - | 5 | 1219 | 40025986 | ||

ZNF699
Genomic Map