siRNA Details
| Gene Symbol: ZNF699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198535.2 | NP_940937.1 | 6 | 984 | ||
| GenBank mRNA | AK095463.1 | - | 4 | 496 | ||
| BC109267.1 | - | 5 | 614 | 40025983 | ||
| BC109268.1 | - | 5 | 614 | 40025986 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF699| Gene Symbol: ZNF699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198535.2 | NP_940937.1 | 6 | 984 | ||
| GenBank mRNA | AK095463.1 | - | 4 | 496 | ||
| BC109267.1 | - | 5 | 614 | 40025983 | ||
| BC109268.1 | - | 5 | 614 | 40025986 | ||