siRNA Details
| Gene Symbol: SBSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166034.1 | NP_001159506.1 | 1 | 381 | ||
| NM_001166035.1 | NP_001159507.1 | 1 | 381 | |||
| NM_198538.3 | NP_940940.1 | 1 | 381 | |||
| XM_011526929.2 | XP_011525231.1 | 1 | 401 | |||
| XM_011526930.2 | XP_011525232.1 | 1 | 357 | |||
| XM_011526931.2 | XP_011525233.1 | 1 | 401 | |||
| GenBank mRNA | AA055367.1 | - | 1 | 167 | ||
| AK291305.1 | - | 1 | 341 | |||
| AY358701.1 | - | 1 | 381 | |||
| BC063640.1 | - | 1 | 323 | 4750640 | ||
| BX648076.1 | - | 1 | 343 | |||

SBSN
Genomic Map