siRNA Details
| Gene Symbol: ZNF829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037232.3 | NP_001032309.2 | 6 | 836 | ||
| NM_001171979.1 | NP_001165450.1 | 6 | 781 | |||
| XM_005258876.4 | XP_005258933.1 | 6 | 659 | |||
| XM_011526933.2 | XP_011525235.1 | 6 | 678 | |||
| GenBank mRNA | AK130969.1 | - | 5 | 516 | ||
| BC107131.1 | - | 6 | 723 | 40025539 | ||
| BC107132.1 | - | 6 | 727 | 40025541 | ||
| BX538288.1 | - | 6 | 782 | |||

ZNF829
Genomic Map