siRNA Details
| Gene Symbol: ZNF568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204835.1 | NP_001191764.1 | 7 | 2316 | ||
| NM_001204836.1 | NP_001191765.1 | 6 | 2058 | |||
| NM_001204837.1 | NP_001191766.1 | 5 | 1999 | |||
| NM_198539.3 | NP_940941.2 | 7 | 2319 | |||
| XM_017026773.1 | XP_016882262.1 | 8 | 2403 | |||
| XM_017026774.1 | XP_016882263.1 | 7 | 2008 | |||
| XM_017026779.1 | XP_016882268.1 | 7 | 2947 | |||
| XM_017026780.1 | XP_016882269.1 | 6 | 2871 | |||
| GenBank mRNA | BX640681.1 | - | 5 | 1983 | ||

ZNF568
Genomic Map