siRNA Details
| Gene Symbol: ZNF773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304334.1 | NP_001291263.1 | 4 | 694 | ||
| NM_001304336.1 | NP_001291265.1 | 4 | 690 | |||
| NM_198542.2 | NP_940944.1 | 4 | 697 | |||
| GenBank mRNA | AK057209.1 | - | 4 | 613 | ||
| BC005167.2 | - | 4 | 610 | 3535599 | ||

ZNF773
Genomic Map