siRNA Details
Gene Symbol: ZNF773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304334.1 | NP_001291263.1 | 4 | 1260 | ||
NM_001304336.1 | NP_001291265.1 | 4 | 1256 | |||
NM_198542.2 | NP_940944.1 | 4 | 1263 | |||
GenBank mRNA | AK057209.1 | - | 4 | 1179 | ||
BC005167.2 | - | 4 | 1176 | 3535599 |
