siRNA Details
| Gene Symbol: DRAXIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198545.3 | NP_940947.3 | 2 | 256 | ||
| XM_011541405.2 | XP_011539707.1 | 3 | 430 | |||
| XM_017001214.1 | XP_016856703.1 | 3 | 566 | |||
| XM_017001215.1 | XP_016856704.1 | 3 | 430 | |||
| GenBank mRNA | AY358750.1 | - | 2 | 217 | ||
| BC111064.1 | - | 2 | 258 | 5769984 | ||
| BM928129.1 | - | 276 | ||||

DRAXIN
Genomic Map