siRNA Details
| Gene Symbol: DRAXIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198545.3 | NP_940947.3 | 5 | 915 | ||
| XM_011541405.2 | XP_011539707.1 | 6 | 1089 | |||
| XM_017001214.1 | XP_016856703.1 | 6 | 1225 | |||
| GenBank mRNA | AB301919.1 | - | 4 | 775 | ||
| AK075558.1 | - | 5 | 910 | |||
| AK311439.1 | - | 2 | 63 | |||
| AY358750.1 | - | 5 | 876 | |||
| BC021286.2 | - | 3 | 265 | |||
| BC111064.1 | - | 5 | 917 | 5769984 | ||

DRAXIN
Genomic Map