siRNA Details
| Gene Symbol: FAM89A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198552.2 | NP_940954.1 | 2 | 457 | ||
| GenBank mRNA | BC009447.1 | - | 2 | 447 | 3530481 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM89A| Gene Symbol: FAM89A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198552.2 | NP_940954.1 | 2 | 457 | ||
| GenBank mRNA | BC009447.1 | - | 2 | 447 | 3530481 | |