siRNA Details
| Gene Symbol: LOC107987394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017030163.1 | XP_016885652.1 | 3, 4 | 323 | ||
| Gene Symbol: LOC100996415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136570.1 | 3, 4 | 179 | |||
| GenBank mRNA | DB092076.1 | - | 3, 4 | 179 | ||

LOC107987394
Genomic Map