siRNA Details
| Gene Symbol: LOC729461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136569.1 | 8 | 979 | |||
| GenBank mRNA | AK129567.1 | - | 8 | 979 | ||
| Gene Symbol: LOC642643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136561.1 | 8 | 979 | |||

LOC729461
Genomic Map