siRNA Details
Gene Symbol: FAM230B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128837.1 | - | 4 | 2616 |
Gene Symbol: LOC100996335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC121069.2 | - | 3 | 417 |
Gene Symbol: LOC642643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136561.1 | 8 | 658 |
Gene Symbol: LOC729461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136569.1 | 8 | 658 | |||
GenBank mRNA | AK129567.1 | - | 8 | 658 |
