siRNA Details
| Gene Symbol: CATIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320865.1 | NP_001307794.1 | 3 | 303 | ||
| NM_198559.1 | NP_940961.1 | 3 | 270 | |||
| XM_005246539.4 | XP_005246596.1 | 2 | 267 | |||
| XM_011511148.2 | XP_011509450.1 | 3 | 402 | |||
| XM_011511149.2 | XP_011509451.1 | 1 | 14 | |||
| XM_017004053.1 | XP_016859542.1 | 1 | 438 | |||
| GenBank mRNA | BC052750.1 | - | 3 | 270 | 5742766 | |

CATIP
Genomic Map