siRNA Details
| Gene Symbol: RBM44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080504.2 | NP_001073973.2 | 13 | 2890 | ||
| XM_005246075.1 | XP_005246132.1 | 13 | 3231 | |||
| XM_011511155.1 | XP_011509457.1 | 5 | 399 | |||
| XM_017004054.1 | XP_016859543.1 | 13 | 3234 | |||
| XM_017004055.1 | XP_016859544.1 | 13 | 3604 | |||
| XM_017004056.1 | XP_016859545.1 | 13 | 3174 | |||
| XM_017004058.1 | XP_016859547.1 | 8 | 1572 | |||
| GenBank mRNA | AK130316.1 | - | 4 | 329 | ||
| BC126111.1 | - | 10 | 973 | |||
| BX648107.1 | - | 12 | 3543 | |||

RBM44
Genomic Map