siRNA Details
| Gene Symbol: ENHO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198573.2 | NP_940975.2 | 2 | 896 | ||
| GenBank mRNA | AY358658.1 | - | 1 | 624 | ||
| BC022101.1 | - | 3 | 706 | 4871461 | ||
| BQ932712.1 | - | 2 | 747 | |||
| BX111579.1 | - | 1 | 483 | |||

ENHO
Genomic Map