siRNA Details
| Gene Symbol: ERCC6L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010895.2 | NP_001010895.1 | 5 | 1263 | ||
| NM_020207.4 | NP_064592.2 | 5 | 1263 | |||
| XM_011518641.2 | XP_011516943.1 | 5 | 1263 | |||
| XM_011518644.2 | XP_011516946.1 | 4 | 1055 | |||
| XM_011518645.2 | XP_011516947.1 | 5 | 1263 | |||
| XM_011518646.2 | XP_011516948.1 | 5 | 1263 | |||
| XM_011518647.1 | XP_011516949.1 | 5 | 1263 | |||
| XM_011518650.2 | XP_011516952.1 | 4 | 415 | |||
| XM_017014707.1 | XP_016870196.1 | 4 | 1055 | |||
| GenBank mRNA | AK307504.1 | - | 4 | 715 | ||
| AK308409.1 | - | 5 | 1195 | |||
| AL389953.1 | - | 5 | 1179 | |||
| BC035183.1 | - | 6 | 1887 | 5266310 | ||
| BC075860.1 | - | 4 | 504 | |||
| BC140702.1 | - | 5 | 1006 | 9021570 | ||
| BC144261.1 | - | 4 | 812 | |||

ERCC6L2
Genomic Map