siRNA Details
| Gene Symbol: ZC3H6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198581.2 | NP_940983.2 | 10 | 2182 | ||
| XM_006712519.2 | XP_006712582.1 | 10 | 2189 | |||
| GenBank mRNA | AB111887.1 | - | 9 | 1628 | ||
| AK127776.1 | - | 7 | 1251 | |||
| AK131416.1 | - | 10 | 2168 | |||
| BC155540.1 | - | 9 | 1628 | |||
| BC167154.1 | - | 9 | 1628 | |||

ZC3H6
Genomic Map