siRNA Details
| Gene Symbol: ZC3H6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198581.2 | NP_940983.2 | 3, 4 | 738 | ||
| XM_006712519.2 | XP_006712582.1 | 3, 4 | 745 | |||
| GenBank mRNA | AB111887.1 | - | 2, 3 | 184 | ||
| AK131416.1 | - | 3, 4 | 724 | |||
| BC155540.1 | - | 2, 3 | 184 | |||
| BC167154.1 | - | 2, 3 | 184 | |||

ZC3H6
Genomic Map