siRNA Details
Gene Symbol: KLHL30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198582.3 | NP_940984.3 | 8 | 1649 | ||
GenBank mRNA | AK125364.1 | - | 7 | 1521 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KLHL30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198582.3 | NP_940984.3 | 8 | 1649 | ||
GenBank mRNA | AK125364.1 | - | 7 | 1521 |