siRNA Details
| Gene Symbol: TRIM74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317815.1 | NP_001304744.1 | 2 | 218 | ||
| NM_198853.2 | NP_942150.1 | 2 | 279 | |||
| XM_011516187.2 | XP_011514489.1 | 2 | 187 | |||
| XM_011516189.2 | XP_011514491.1 | 2 | 503 | |||
| GenBank mRNA | AF498999.1 | - | 2 | 279 | ||
| BC033871.1 | - | 2 | 218 | 5201777 | ||
| Gene Symbol: TRIM73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198924.3 | NP_944606.2 | 2 | 279 | ||
| GenBank mRNA | AF498998.1 | - | 2 | 279 | ||
| DA452220.1 | - | 2 | 152 | |||
 Back To Top
 Back To Top
 TRIM74
TRIM74 Genomic Map
Genomic Map Back To Top
 Back To Top