siRNA Details
| Gene Symbol: KCTD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198404.2 | NP_940686.2 | 2 | 856 | ||
| GenBank mRNA | AK290675.1 | - | 2 | 858 | ||
| BC018063.1 | - | 1 | 602 | 4814899 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCTD4| Gene Symbol: KCTD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198404.2 | NP_940686.2 | 2 | 856 | ||
| GenBank mRNA | AK290675.1 | - | 2 | 858 | ||
| BC018063.1 | - | 1 | 602 | 4814899 | ||