siRNA Details
| Gene Symbol: KRTAP10-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ566381.1 | - | 1 | 199 | ||
| Gene Symbol: KRTAP10-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198690.2 | NP_941963.2 | 1 | 1011 | ||
| GenBank mRNA | AB076357.1 | - | 2 | 293 | ||

KRTAP10-2
Genomic Map