siRNA Details
| Gene Symbol: SOGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012279.2 | NP_001012279.1 | 6 | 2477 | ||
| GenBank mRNA | AK091416.1 | - | 3 | 248 | ||
| AK096490.1 | - | 5 | 922 | |||
| BC127760.1 | - | 5 | 746 | |||
| BC143043.1 | - | 5 | 746 | |||
| BC143044.1 | - | 5 | 746 | |||

SOGA3
Genomic Map