siRNA Details
| Gene Symbol: CEP85L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042475.2 | NP_001035940.1 | 3 | 1347 | ||
| NM_001178035.1 | NP_001171506.1 | 4 | 835 | |||
| NM_206921.2 | NP_996804.2 | 3 | 1347 | |||
| XM_005266970.1 | XP_005267027.1 | 3 | 867 | |||
| XM_005266971.1 | XP_005267028.1 | 3 | 830 | |||
| XM_006715475.3 | XP_006715538.1 | 4 | 1043 | |||
| XM_011535809.2 | XP_011534111.1 | 4 | 1938 | |||
| XM_011535810.2 | XP_011534112.1 | 5 | 972 | |||
| XM_011535811.1 | XP_011534113.1 | 3 | 733 | |||
| XM_017010846.1 | XP_016866335.1 | 5 | 971 | |||
| XM_017010847.1 | XP_016866336.1 | 3 | 1162 | |||

CEP85L
Genomic Map