siRNA Details
| Gene Symbol: ZNF322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242797.1 | NP_001229726.1 | 5 | 1229 | ||
| NM_001242798.1 | NP_001229727.1 | 3 | 1084 | |||
| NM_001242799.1 | NP_001229728.1 | 3 | 1065 | |||
| NM_024639.4 | NP_078915.2 | 4 | 1154 | |||
| GenBank mRNA | AK027046.1 | - | 1 | 474 | ||
| AK290038.1 | - | 4 | 879 | |||
| AY376736.1 | - | 4 | 937 | |||
| BC050425.1 | - | 1 | 650 | 6473688 | ||
| BC074772.2 | - | 1 | 446 | |||
| BC119669.2 | - | 1 | 567 | 40116184 | ||

ZNF322
Genomic Map