siRNA Details
| Gene Symbol: GPR153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207370.2 | NP_997253.2 | 3 | 1011 | ||
| XM_011541434.2 | XP_011539736.1 | 3 | 1115 | |||
| XM_017001250.1 | XP_016856739.1 | 2 | 1097 | |||
| XM_017001251.1 | XP_016856740.1 | 3 | 1000 | |||
| GenBank mRNA | AY255529.1 | - | 1 | 274 | ||
| BC068275.1 | - | 3 | 1011 | 5167285 | ||

GPR153
Genomic Map