siRNA Details
| Gene Symbol: SLC16A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213606.3 | NP_998771.3 | 8 | 1792 | ||
| XM_017016237.1 | XP_016871726.1 | 7 | 1882 | |||
| XM_017016238.1 | XP_016871727.1 | 7 | 1821 | |||
| XM_017016239.1 | XP_016871728.1 | 7 | 1680 | |||
| GenBank mRNA | AK124901.1 | - | 4 | 1071 | ||
| AK127303.1 | - | 8 | 1793 | |||
| BC086873.1 | - | 3 | 745 | |||
| CR936657.1 | - | 6 | 3990 | |||

SLC16A12
Genomic Map