siRNA Details
| Gene Symbol: CC2D2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001732.3 | NP_001001732.2 | 7 | 814 | ||
| NM_001159747.1 | NP_001153219.1 | 8 | 847 | |||
| XM_011539789.2 | XP_011538091.1 | 15 | 1908 | |||
| XM_011539790.2 | XP_011538092.1 | 14 | 1794 | |||
| XM_011539791.2 | XP_011538093.1 | 14 | 1725 | |||
| XM_011539792.2 | XP_011538094.1 | 9 | 1030 | |||
| XM_011539793.2 | XP_011538095.1 | 8 | 2009 | |||
| XM_011539795.2 | XP_011538097.1 | 7 | 1826 | |||
| XM_017016240.1 | XP_016871729.1 | 8 | 1086 | |||
| GenBank mRNA | AK302377.1 | - | 8 | 826 | ||
| BC075861.1 | - | 7 | 779 | 5171414 | ||

CC2D2B
Genomic Map