siRNA Details
Gene Symbol: OR56B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005180.2 | NP_001005180.1 | 1 | 489 | ||
GenBank mRNA | AK122668.1 | - | 1 | 489 | ||
BC137184.1 | - | 1 | 486 | 9021181 | ||
BC137185.1 | - | 1 | 486 | 9021182 |
